Learn More
ATCC Primary Lung Smooth Muscle Cells; Normal, Human
Supplier:  ATCC PCS130010
Encompass
Primary Lung Smooth Muscle Cells are myocytes isolated from the lung of a donor. The cell can be used for the study of angiogenesis, atherosclerosis, diabetes, or vascular/pulmonary biology.
Catalog No. 50-238-5086
 Restricted Item