Learn More
ATCC Primary Mammary Epithelial Cells Normal Human
Supplier:  ATCC PCS600010
Encompass
ATCC's Human Mammary Epithelial Cells cultured in Mammary Epithelial Cell complete medium (basal medium plus growth kit) is an optimal serum-free culture model for many research areas. Common uses of HMEC include the study of breast cancer development, 3-dimensional culture and carcinogen screening.
Catalog No. 50-238-2639