Learn More
ATCC Primary Prostate Epithelial Cells; Normal Human
Supplier:  ATCC PCS440010
Encompass
Normal human primary prostate epithelial cells were isolated from the prostate of a donor and can be used in hormonal regulation of the prostate, regulation control of the secretory function of prostate cells, and as a control for the study of prostate cancer.
Catalog No. 50-238-1932