Learn More
ATCC Primary Renal Proximal Tubule Epithelial Cells; Normal, Human
Supplier:  ATCC PCS400010
Encompass
Primary Renal Proximal Tubule Epithelial Cells; Normal, Human (RPTEC) provide an ideal cell system to propagate renal epithelial cells in low serum (0.5% FBS) conditions when grown in Renal Epithelial Cell Basal Media supplemented with Renal Epithelial Cell Growth Kit components. The cells are cryopreserved in the second passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements and reagents are quality tested together to guarantee optimum performance and reliability.
Catalog No. 50-238-2398