Learn More
ATCC Primary Umbilical Cord-Derived Mesenchymal Stem Cells; Normal, Human
Supplier:  ATCC PCS500010
Encompass
Normal Human Umbilical Cord-Derived Mesenchymal Stem Cells, when grown in Mesenchymal Stem Cell Basal Media supplemented with Mesenchymal Stem Cell Growth Kit for Adipose and Umbilical-derived MSCs - Low serum components, provide an ideal cell system to propagate mesenchymal stem cells in low serum (2% FBS) conditions. When maintained under optimal growth conditions, ATCC Normal Human Umbilical Cord-Derived Mesenchymal Stem Cells have been shown to be multipotent, capable of differentiating down the adipogenic, osteogenic, and chondrogenic lineages. The cells are cryopreserved at the second passage to ensure the highest viability and plating efficiency. ATCC Primary Cell Solutions cells, media, supplements, and reagents are quality tested together to guarantee optimum performance and reliability.
Catalog No. 50-238-2295