Learn More
ATCC Primary Uterine; Fibroblast Cells; Normal, Human
Supplier:  ATCC PCS460010
Encompass
Primary uterine fibroblast cells are spindle-shaped and were isolated from the uterus of a donor, and can be used in female reproductive biology, drug testing, and oncology studies.
Catalog No. 50-238-2347