Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV FreezIS, 100 mL
Supplier:  FUJIFILM BIOSCIENCES INC 91139100ML/DEL
Encompass_Preferred
Enables cell preservation for short-term at -80°C and long-term storage in liquid nitrogen to -196°CHigh post-thaw viability and growthMaintains cell surface marker expression of mesenchymal stem/ stromal cells (MSCs) and PBMCs (T cells) and hematopoietic stem cells (HSCs) post-thawComplete, ready-to-use mediumContains DMSO in the formulationBy purchasing this product you are agree that you will not use this product as excipient
Catalog No. NC1064100
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product