Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV Human Fibronectin1mg
Supplier:  FUJIFILM BIOSCIENCES INC 310021MG
Encompass_Preferred
PRIME-XV Human Fibronectin can be used as a coating cell culture substrate to support optimal attachment and growthof human primary cells, including stem/ progenitor cells. The recommended concentration for this application is typically 1- 5 μg/ mL. It can also be added to the media to support cell spreading at a concentration of 0.5-50 μg/ mL. PRIME-XV Human Fibronectin is derived from human plasma. The human serum used for the purification of this product was certifiedby the supplier to be HIV and HBsAg negative at the time of shipment. Human blood products should always be treated inaccordance with universal handling precautions. PRIME-XV Human Fibronectin is supplied as 0.2 μm filtered solution inTris-HCl, NaCl and Urea
Catalog No. 50-271-7119