Learn More
FUJIFILM BIOSCIENCES INC PRIME XV IS21 SUPPLEMENT (50X)
Supplier:  FUJIFILM BIOSCIENCES INC 9114210ML/DEL
Encompass_Preferred
PRIME-XV IS21 Supplement (50X) is a serum-free supplement optimized for use with PRIME-XV Neural Basal Medium to support maintenance and long-term culture of neuronal cells. This supplement is supplied as a 50X concentrated solution and should be diluted to 1X in basal medium before use. This supplement does not contain antibiotics.
Catalog No. NC1404555
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.