Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV NK Cell CDM, 1L
Supplier:  FUJIFILM BIOSCIENCES INC 912151L/DEL
Encompass_Preferred
Designed for use in NK and NKT cell-based immunotherapy research and translational applications, this advanced formulation delivers growth while maintaining NK and NKT cell functionality and potency - without the need for serum supplementation.Provides Optimal PerformanceChemically defined, animal component-free natural killer cell culture mediumSupports robust cell expansion and viability in heterogeneous PBMC culture enriched for CD56+ cellsSupports expansion of NK cells when supplemented with IL-2, IL-12, IL-18, and IL-21Supports NKT cell expansion when supplemented with IL-2Maintains high NK and NKT cytotoxic capabilityDesigned and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management
Catalog No. NC1698646
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.