Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV T Cell CDM, 1 L
Supplier:  FUJIFILM BIOSCIENCES INC 911541L/DEL/DEL
Encompass_Preferred
PRIME-XV T Cell CDM is the first commercially-available, chemically defined, animal component-free medium for the expansion and transduction of human T cells. The formulation is optimized to deliver consistently vigorous growth while maintaining T cell functionality and potency.Supports T cell expansion in static and dynamic automation systems while maintaining functionalityEliminates the adverse effects undefined components cause on T cell phenotypesSupports polarization to targeted T cell types, such as Th1, Th2, cytotoxic T cells, to further possible therapy applicationsPerforms transduction of T cells without requiring spinoculation or transduction enhancers
Catalog No. NC1275575
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product