Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV T Cell Expansion XSFM, 1 L
Supplier:  FUJIFILM BIOSCIENCES INC 911411L/DEL
Encompass_Preferred
Optimized for activation and expansion of human T lymphocytesMaintains potency of major T cells subsets such as, CD4+ and CD8+Supports expansion of re-activated T cellsSerum-free and xeno-free formulation reduces risk of contamination and provides lot-to-lot consistencyContains gentamicin for reduced contamination of culture componentsDesigned and manufactured to facilitate transfer from research to clinic. Supported by robust raw material controls and supply chain management.
Catalog No. NC1123070