Learn More
DSI LLC PRIMESEP SB 4.6 X150 MM
Supplier:  DSI LLC SB46.150.0510
Encompass_Preferred
A reverse-phase analytical column with strong embedded basic ion-pairing groups. Improves retention of acidic compounds by anion-exchange mechanism. Separates bases by ion-exclusion. Retains neutral compounds by reverse-phase.
Catalog No. NC0543305