Learn More
Sielc Technologies Inc PRIMESEP SB HPLC COLOMN
Supplier:  Sielc Technologies Inc SB32.150.0510
Encompass
Primesep SB HPLC Colomn 150mm length 3.2 mm diam 5um particle size pore size 100a standard hardware
Catalog No. NC1619140