Learn More
Clontech Labs 3P PrimeSTAR® GXL DNA Polymerase, 1,000 Units
Supplier: Clontech Labs 3P R050B
A hot-start, high-fidelity PCR enzyme, PrimeSTAR GXL DNA Polymerase excels in reactions with GC-rich templates, excess template, and long amplicons up to 30 kb (GXL). The polymerase is supplied with separate tubes of optimized buffer (Mg2+ plus), and dNTPs. 1,000 Units
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.