Learn More
Integrated DNA Technologies Inc PrimeTime Gene Expression Master Mix, 25mL
Supplier:  Integrated DNA Technologies Inc 1055771
Encompass
Ready-to-use, 2X concentrated master mix that contains an antibody-mediated, hot-start DNA polymerase; dNTPs; MgCl2; enhancers; and stabilizers. Individual tubes of reference dye included.
Catalog No. NC1666379
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.