Learn More
FUJIFILM BIOSCIENCES INC PRIME-XV Stem FreezIS DMSO-Free, 10 mL
Supplier:  FUJIFILM BIOSCIENCES INC 9114010ML/DEL
Encompass_Preferred
Eliminate the risk of DMSO toxicity and maintain potency of human mesenchymal stem/ stromal cells (MSCs), PBMCs (T cells), and hematopoietic stem cells (HSCs) throughout cryopreservationComparable post-thaw cell viability to solutions containing DMSONontoxic when injected in animal models (refer to toxicological application note)Enables cell preservation for short-term at -80°C* and long-term storage in liquid nitrogen to -196°CComplete, ready-to-use medium* Human MSC and PBMC (T cell) data available for short-term storage. Human HSC data is not available.By purchasing this product you are agree that you will not use this product as excipient
Catalog No. NC1612461
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product