Learn More
Medchemexpress LLC Prinomastat hydrochloride | 1435779-45-5 | 98.2% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY12170A5MG
Encompass_Preferred
Prinomastat hydrochloride is the hydrochloride salt of prinomastat (AG3340), a potent, orally active broad-spectrum matrix metalloproteinase (MMP) inhibitor used in preclinical in vitro and in vivo research. Supplied as a solid powder, it is intended for laboratory research use.
  • Potent broad-spectrum MMP inhibitor.
  • Hydrochloride salt supplied as a solid powder.
  • Molecular formula C18H22ClN3O5S2 and molecular weight 459.97 g/mol.
  • High purity suitable for research, reported at approximately 98.2%.
  • Can be prepared as concentrated stock solutions; aqueous stocks may require sonication.
Catalog No. 50-002-30804