Learn More
Leica Biosystems Leica Print Head Cleaning Pens (Pack of 5) for Slide Printers
Supplier:  Leica Biosystems 14060655774
Encompass_Preferred
Leica Biosystems print head cleaning pens are a pack of five maintenance pens designed to remove residue and debris from thermal print heads and help maintain consistent print quality for slide and laboratory printers that use thermal printing technology.
  • Pack of five pens for multiple maintenance cycles.
  • Designed to clean thermal print heads and remove residue.
  • Helps reduce streaking and improve print quality.
  • Easy-to-use pen format for precise cleaning of print elements.
  • Recommended for routine maintenance of slide and laboratory printers.
Catalog No. NC3360614