Learn More
Bioworld PRIONEX--100 ML
Supplier:  Bioworld 416201682
Encompass_Preferred
Prionex, 100mL, pH: 6.0 to 7.5, Freely soluble in water, dilute electrolyte solutions, glycerol and DMSO
Catalog No. NC9183395