Learn More
Medchemexpress LLC Pristanic acid | 1189-37-3 | 99.8% | 298.50 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11302410MM/1ML
Encompass_Preferred
Pristanic acid is an endogenous metabolite present in blood. It is used for the research of Alpha Methylacyl CoA Racemase Deficiency and Zellweger Syndrome.
  • Endogenous metabolite
  • Used for the research of Alpha Methylacyl CoA Racemase Deficiency and Zellweger Syndrome
Catalog No. 50-004-39771