Learn More
Cayman Chemical PrIstImerIn 10mg
SDP
Supplier:  Cayman Chemical 1362110 MG
Encompass_Preferred
A naturally occuring terpenoid that potently inhibits MAGL (IC50 = 93 nM); at 1 µM, significantly inhibits endogenous MAGL in isolated rat neurons
Catalog No. 50-295-3341