Learn More
BIRCH BIOTECH LLC PRISTINE Tetrahydrofuran, HPLC Grade, 4 Liter, 4/case
Supplier:  BIRCH BIOTECH LLC 19175
Encompass_Preferred
PRISTINE Tetrahydrofuran, HPLC Grade, 4 Liter, 4/caseHigh-Purity Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Tetrahydrofuran (THF) is a high-purity solvent designed for high-performance liquid chromatography and other sensitive analytical applications. With its low levels of impurities and low UV absorbance, this THF grade ensures minimal background interference, supporting high accuracy in both qualitative and quantitative analyses. Ideal for applications requiring precise separation and detection, it provides consistent results critical to advanced research and industrial applications.
Catalog No. NC3861159