Learn More
Medchemexpress LLC Pritelivir | 348086-71-5 | C18H18N4O3S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15303100MG
Encompass_Preferred
Pritelivir (AIC316) is an antiviral agent that functions as an inhibitor of the viral helicase-primase complex. This mechanism of action allows it to exhibit antiviral activity both in laboratory settings (in vitro) and in animal models of herpes simplex virus (HSV) infection. It is specifically active against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2).
  • Inhibits the viral helicase-primase complex.
  • Demonstrates antiviral activity in vitro and in animal models of HSV infection.
  • Effective against herpes simplex virus types 1 and 2 (HSV-1 and HSV-2).
  • Exhibits an IC50 of 0.02 μM against HSV1-2.
Catalog No. 50-003-73170