Learn More
Selleck Chemical LLC Pritelivir
SDP
Supplier:  Selleck Chemical LLC S75465MG
Encompass_Preferred
Pritelivir (BAY 57-1293 AIC316) is a potent helicase primase inhibitor exhibiting antiviral effect on herpes simplex virus (HSV) with IC50 of 20 nM for both HSV-1 and HSV-2
Catalog No. 50-000-00074