Learn More
Medchemexpress LLC Pritelivir mesylate hydrate | 1428321-10-1 | 99.5% | 516.61 g/mol | C19H24N4O7S3 | 10 MG

Supplier: Medchemexpress LLC HY15303B10MG
Pritelivir mesylate hydrate is the mesylate hydrate salt of pritelivir, a research-grade antiviral inhibitor of the viral helicase-primase complex. Supplied as a solid for in vitro and preclinical research, it exhibits potent activity against herpes simplex virus and is provided with analytical data for identity and purity.
- Potent inhibitor of the viral helicase-primase complex.
- Active against HSV-1 and HSV-2 (reported IC50 ~0.02 μM).
- High purity solid (≈99.5%).
- Molecular weight 516.61 g/mol; formula C19H24N4O7S3.
- Provided in small research pack sizes for laboratory use.
- Store sealed at 4°C; in solvent: -80°C (6 months), -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.