Learn More
Cayman Chemical PRL-3 InhIbItr 1mg
SDP
Supplier:  Cayman Chemical 187191 MG
Encompass_Preferred
A cell-permeable benzylidene rhodamine that inhibits PRL-3 (IC50 = 900 nM for human PRL-3 in vitro), with minimal activity against other phosphatases; reduces the invasion of mouse melanoma B16F10 cells in a cell-based assay
Catalog No. 50-298-0322