Learn More
Medchemexpress LLC PRLX-93936 (dihydrochloride) | 1094210-96-4 | 99.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY119264A5MG
Encompass_Preferred
PRLX-93936 dihydrochloride is a research-grade HIF-1α inhibitor supplied as the dihydrochloride salt. It is provided as a white to off-white solid in milligram-scale quantities for cellular and biochemical studies of hypoxia signaling and preclinical oncology research.
  • High purity: 99.8% typical, suitable for research applications.
  • Salt form: dihydrochloride for improved stability and solubility handling.
  • Small-scale packaging: available in milligram quantities for screening and early-stage studies.
  • Physical form: white to off-white solid, convenient for weighing and formulation.
  • Includes supporting documentation: product datasheet and safety data sheet available.
Catalog No. 50-002-33065