Learn More
Cayman Chemical PRMT4CARM1 InhIbItr 500ug
SDP
Supplier:  Cayman Chemical 11033500 UG
Encompass_Preferred
A selective inhibitor of PRMT4/CARM1 (IC50 = 7.1 µM); reduces the prostate-specific antigen promoter activity in human prostate cancer LNCaP cells
Catalog No. 50-295-1089