Learn More
Cayman Chemical PRMT4CARM1 InhIbItr 5mg
SDP
Supplier:  Cayman Chemical 110335 MG
Encompass_Preferred
A selective inhibitor of PRMT4/CARM1 (IC50 = 7.1 µM); reduces the prostate-specific antigen promoter activity in human prostate cancer LNCaP cells
Catalog No. 50-295-1364