Learn More
GA International DYMO LabelWriter™ 550 Printing Kit: LabelWriter™ 550 Label Printer + Cryo Label (x2) LWCS-505-WH
Supplier:  GA International PKTLWCS505NA
Encompass_Preferred
Our DYMO LabelWriter™ 550 printing kit provides an affordable, complete printing solution for labs on a budget. Supplied with the DYMO LabelWriter™ 550 direct thermal printer, free DYMO Connect label design/barcoding software, and two rolls of LWCS-class labels, this kit provides you with all you need to start printing labels for your lab.Features:- Small, lightweight direct thermal printer, eliminating the need for ink or toner - The direct thermal printing technology adjusts to the loaded DYMO-branded labels, producing crisp barcodes, images, and text - Easy-to-use DYMO Connect software (included) is compatible with either Windows 8.1 (or later) and Mac 10.14 (or later) - LWCS-class labels are suitable for quick labeling of frozen vials and tubes (no thawing required) What’s included in the Printing Kit:- LabelWriter™ 550 Label Printer- Starter roll of address labels- Two rolls of LWCS-505-WH- Power Adapter- Power Cable- USB Cable- Quick Start Guide
Catalog No. NC3454633