Learn More
GA International PRNTNGKT-LWCS-506-NA-1KT
Supplier:  GA International PKTLWCS506NA
Encompass_Preferred
  • Comprehensive direct thermal labeling solution for scientific laboratories.
  • Designed specifically for cryogenic applications.
  • Includes a DYMO LabelWriter 550 printer.
  • Offers DYMO Connect label design software for barcodes, text, and graphics.
  • No ink or toner is required.
  • Compatible with Windows 8.1, Windows 10, Mac 10.14, and macOS 13.
  • Features CryoSTUCK with SnapPEEL™ label technology.
  • Labels are resistant to cryogenic temperatures and solvents.
  • Suitable for frozen surfaces without thawing samples.
  • Durable solvent-resistant labels maintain integrity in ultra-low temperatures.
  • Each roll contains 750 labels.
  • Label dimensions: 0.94 in x 0.5 in + 0.375 in circle.
  • Minimum label application temperature: -80 °C.
Catalog No. NC3454635