Learn More
CPC Scientific H-Ala-Arg-Leu-Asp-Val-Ala-Ser-Glu-Phe-Arg-Lys-Lys-Trp-Asn-Lys-Trp-Ala-Leu-Ser-Arg-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM007B
Encompass
Precursor of adrenomedullin containing proadrenomedullin NH2-terminal 20 peptide (PAMP), a unique 20-residue sequence. When administered, PAMP produces a strong and rapid hypotensive effect in anesthetized rats. PAMP has also been suggested to function as an inhibitory modulator of renal noradrenergic neurotransmission, thereby playing an important role in the regulation of renal functions. PAMP has been proposed to serve as an inhibitory regulator in adrenal catecholamine release in vivo and possesses a potent hyperglycemic effect after intra-third cerebroventricular administration in fasted mice. ONE-LETTER SEQUENCE: ARLDVASEFRKKWNKWALSR-NH2MOLECULAR FORMULA: C112H178N36O27MOLECULAR WEIGHT:2460.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [150238-87-2], RESEARCH AREA: CardiovascularREFERENCES: K. Kitamura et al., BBRC, 194, 720 (1993)
Catalog No. 50-210-8628