Learn More
Medchemexpress LLC Pro-Phe-Arg-AMC acetate | 2237216-30-5 | 99.17% | 635.71 | 10 MG

Supplier: Medchemexpress LLC HYP261810MG
Pro-Phe-Arg-AMC acetate is a synthetic peptide that serves as a substrate for human glandular kallikrein 2 (hK2). This compound is typically provided as a white to off-white solid, ensuring high purity for research applications.
- Used as a peptide substrate for human glandular kallikrein 2 (hK2).
- High purity of 99.17%.
- Molecular weight of 635.71.
- Store powder sealed, away from moisture, at -80°C for 2 years or -20°C for 1 year.
- Store in solvent sealed, away from moisture, at -80°C for 6 months or -20°C for 1 month.
- Soluble in DMSO at 100 mg/mL (157.30 mM); requires ultrasonic.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.