Learn More
Medchemexpress LLC Proanthocyanidins | 20347-71-1 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN079450MG
Encompass_Preferred
Proanthocyanidin (Procyanidin) is a class of polyphenolic compounds widely distributed in higher plants, consisting of an electrophilic flavanyl unit. It can be used as an antioxidant and anti-cancer agent. Additionally, Proanthocyanidin exhibits anti-inflammatory, cardioprotective, antibacterial, and antifungal properties, making it useful in treating chronic venous insufficiency, capillary fragility, sunburn, and retinopathy.
  • Antioxidant
  • Anti-cancer agent
  • Anti-inflammatory
  • Cardioprotective
  • Antibacterial
  • Antifungal properties
  • Can inhibit P-fimbriated *E. coli* from adhering to uroepithelial cells
  • Shows strong antimutagenic effects
  • Inhibits rat pancreatic carcinogenesis in the initiation stage
Catalog No. 50-004-16913