Learn More
Hach Company Electrode Cleaning Solution for Fats, Oils and Grease Samples, 500 mL
Supplier:  Hach Company 2964449
Encompass
Electrode cleaning solution to remove fats, oils and grease contaminations/deposits from pH sensing element. 500 mL bottle
Catalog No. 50-199-6956