Learn More
Selleck Chemical LLC Probenecid 10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S402210MM/1ML
Encompass_Preferred
Probenecid (Benemid) is a classical competitive inhibitor of organic anion transport, which is also a TRPV2 agonist and an inhibitor of TAS2R16. Probenecid is also a clinically used broad-spectrum Pannexin1 (Panx1) blocker. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3130