Learn More
Abcam Probenecid, ABC-transporter inhibitor, 1G
Supplier:  Abcam AB1457251G
Encompass_Preferred
MW 285.36 Da, Purity >98%. ABC-transporter inhibitor. Competitive MRP1 blocker. Pannexin channel blocker (IC₅₀ = 150 μM). Can be used to block the efflux of intracellular dyes and indicators.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5076