Learn More
Medchemexpress LLC Probenecid sodium | 23795-03-1 | MFCD01661360 | 99.4% | 307.34 | C13H18NNaO4S | 10 G
SDP
Supplier:  Medchemexpress LLC HYW40491610G
Encompass_Preferred
Probenecid sodium is the sodium salt of probenecid supplied for laboratory research. It acts as a TRPV2 agonist and an inhibitor of organic-anion transporters, provided as a white to off-white solid characterized by CAS 23795-03-1 and molecular formula C13H18NNaO4S.
  • Sodium salt form with improved water solubility.
  • Active as a TRPV2 agonist and organic-anion transport inhibitor.
  • High purity (99.4%).
  • Available in research-scale pack sizes, including 10 G.
  • Suitable for analytical and biological assays.
Catalog No. 50-002-81666