Learn More
Medchemexpress LLC Probenecid (sodium) | 23795-03-1 | MFCD01661360 | 99.4% | 307.34 | C13H18NNaO4S | 50 G
SDP
Supplier:  Medchemexpress LLC HYW40491650G
Encompass_Preferred
Probenecid sodium is the sodium salt of probenecid supplied as a solid for research use. It is reported to act as a selective transient receptor potential vanilloid 2 (TRPV2) agonist and to inhibit pannexin 1 channels. The product is provided with high reported purity and is available in laboratory-scale pack sizes.
  • Sodium salt of probenecid as listed on the SDS.
  • Selective TRPV2 agonist and pannexin 1 inhibitor.
  • High reported purity (99.37%).
  • Solid form suitable for handling and storage.
  • Available in laboratory pack sizes, including 50 G.
  • CAS number 23795-03-1 documented for identification.
Catalog No. 50-002-81596