Learn More
Advanced ImmunoChemical, Inc. Human Recombinant PROBNP, Glycosylated, 0.05mg
Supplier: Advanced ImmunoChemical, Inc. 8GPRONP0.05MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Human recombinant proBNP, glycosylated. 0.05mg. Calibrator or standard in immunoassays, immunogen for antibody production Purity > 95% (Tricine-SDS-PAGE in reducing conditions).ProBNP concentration is determined spectrophotometrically Immunoaffinity purification followed by ion-exchange Lyophilized from 300 mM NaCl, 50 mM sodium acetate, pH 4.2.Storage Conditions:Before and after reconstitution at -70 °C (between -65 and -80 °C allowed)Other information:Avoid repeated freezing and thawing. It is recommended to aliquot the product after the first thawing.To avoid loss of immunological activity it is recommended to reconstitute lyophilized protein in 50 mM sodium acetate, pH 4.2 to obtain the same proBNP concentration that was before lyophilization. After reconstitution store at -70 °C.Material safety note:This product is sold for research use only.