Learn More
YMC America, Inc ProC18 120A 2um 5x0.3mm 1/16
SDP
Supplier:  YMC America, Inc AS12S02E5H0AU
Encompass
YMC-Pack Pro C18 columns start with a tightly controlled highly purified silica gel followed by a high-coverage C18 bonding and finished with a unique endcapping procedure utilizing Lewis acid/Lewis base chemistry Together these features provide unequaled separations with sharp and symmetrical peaks for all types of organic molecules especially basic pharmaceuticals and chelating compounds
Catalog No. 50-008-79078