Learn More
YMC America, Inc ProC18 RS WX 80A 5um 250x30mm
SDP
Supplier:  YMC America, Inc RS08S052530WX
Encompass
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds
Catalog No. 50-008-78091