Learn More
YMC America, Inc ProC18RS col 80A 3um 250x4.6mm
SDP
Supplier:  YMC America, Inc RS08S032546WT
Encompass_Preferred
YMC-Pack Pro C18 RS exhibits elevated recognition of stereoselectivity due to high carbon trifunctional bonding Columns maintain a strong resistance to acid and alkali and the separation selectivity for compounds that differ only slightly in hydrophobicity is outstanding In addition extensive endcapping of the stationary phase results in excellent peak shape for basic compounds
Catalog No. 50-008-81809