Learn More
Medchemexpress LLC Procainamide hydrochloride | 614-39-1 | 99.5% | C13H22ClN3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYA008450MG
Encompass_Preferred
Procainamide hydrochloride is a specific and potent inhibitor of DNA methyltransferase 1 (DNMT1). It reactivates the expression of tumor suppressor factors by demethylating tumor suppressor genes, induces vacuolization in various cell types, and reduces cell proliferation and migration. Additionally, it relaxes airway smooth muscle by activating potassium channels, making it applicable in cancer and arrhythmia research.
  • Specific and potent inhibitor of DNA methyltransferase 1 (DNMT1)
  • Reactivates tumor suppressor gene expression
  • Induces vacuolization in various cell types
  • Reduces cell proliferation and migration
  • Relaxes airway smooth muscle through potassium channel activation
  • Applicable in cancer and arrhythmia research
Catalog No. 50-004-32336