Learn More
Apexbio Technology LLC Procaine 59-46-1 5g
SDP
Supplier:  Apexbio Technology LLC B34355000
Encompass_Preferred
Procaine (CAS 59-46-1) is a small-molecule inhibitor targeting voltage-gated sodium channels It is designed to block these channels thereby inhibiting neuronal action potential propagation and suppressing nerve signal transmission Procaine exerts its biological activity primarily through reversible inhibition of voltage-gated sodium channel currents In in vitro studies Procaine demonstrates sodium channel inhibition with reported IC50 values typically ranging from 250 to 400 M depending on the experimental system Based on these pharmacological properties Procaine holds research potential in studies of electrophysiology ion channel pharmacology and neurological signaling pathways
Catalog No. 50-000-94524