Learn More
Medchemexpress LLC Procaine (hydrochloride) | 51-05-8 | 97.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB05461G
Encompass_Preferred
Procaine hydrochloride is a DNA-demethylating agent for research use only, acting through multiple targets with a slow onset and short duration of action. In laboratory settings, it inhibits 5-HT3 receptor-mediated inward current and decreases 5-methylcytosine DNA content. It can also demethylate highly hypermethylated CpG islands, suppress cancer cell growth by inducing mitotic arrest, stimulate limbic system cells, and modify synaptic transmission in amygdala output pathways.
  • Inhibits 5-HT3 receptor-mediated inward current
  • Decreases 5-methylcytosine DNA content
  • Demethylates highly hypermethylated CpG islands
  • Suppresses cancer cell growth by inducing mitotic arrest
  • Stimulates limbic system cells
  • Modifies synaptic transmission in amygdala output pathways
Catalog No. 50-004-08594