Learn More
Selleck Chemical LLC Procaine penicillin G S4979-1g
SDP
Supplier:  Selleck Chemical LLC S49791G
Encompass_Preferred
Penicillin G Procaine (PGP Penicillin G Procaine Bicillin C-R) a crystalline complex combining penicillin G with procaine is a -lactam antibiotic
Catalog No. 50-310-9003