Learn More
TARGETMOL CHEMICALS INC Procaine penicillin G 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T4553100MG
Encompass_Preferred
Also available in 1 mL, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes.Procaine penicillin G (Duphapen) (procaine benzylpenicillin) is an antibiotic useful for the treatment of a number of bacterial infections. Specifically, it is used for syphilis, anthrax, mouth infections, pneumonia, diphtheria, cellulitis, and animal bites. Purity 98.64%
Catalog No. 50-288-0172