Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Procarbazine (Hydrochloride) | 366-70-1 | 99.7% | 257.76 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY13733500MG

Encompass_Preferred

Procarbazine (Hydrochloride) is an orally active alkylating agent with anticancer activity. It is suitable for research applications, particularly in studies related to Hodgkin's disease.

  • Orally active alkylating agent
  • Demonstrates anticancer activity
  • Suitable for Hodgkin's disease research
  • High purity of 99.7%
  • Provided as a solid
  • Recommended storage for solid: 4°C, sealed, away from moisture
  • Recommended storage for in solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-51313


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.